2-[(4-Methylbenzyl)sulfanyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045596-500MG 045596-1G 045596-5G
101871-670EA 176.92 USD
101871-670 101871-672 101871-674
2-[(4-Methylbenzyl)sulfanyl]benzenecarboxylic acid ≥95%
2-[(4-Methylbenzyl)sulfanyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₂S
Melting Pt: 183…187 °C
MDL Number: MFCD01935992
CAS Number: 104351-51-1

Order Now

Specification Test Results

Melting point 183-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR