1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-Indol-3-yl acetic acid ≥95%

Supplier: Matrix Scientific

045590-500MG 045590-1G 045590-5G
101871-640EA 556.92 USD
101871-640 101871-642 101871-644
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-Indol-3-yl acetic acid ≥95%
1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-Indol-3-yl acetic acid

Hazard: Irritant

Melting Pt: 158…160 °C
MDL Number: MFCD00664543
CAS Number: 339099-14-8

Order Now

Specification Test Results

Melting point 158-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR