{2-[(3,4-Dichlorobenzyl)sulfanyl]-1-methyl-1H-imidazol-5-yl}methanol ≥95%

Supplier: Matrix Scientific

045589-500MG 045589-1G 045589-5G
101871-634EA 375.38 USD
101871-634 101871-636 101871-638
{2-[(3,4-Dichlorobenzyl)sulfanyl]-1-methyl-1H-imidazol-5-yl}methanol ≥95%
{2-[(3,4-Dichlorobenzyl)sulfanyl]-1-methyl-1H-imidazol-5-yl}methanol

Hazard: Irritant

Formula: C₁₂H₁₂Cl₂N₂OS
Melting Pt: 127…129 °C
MDL Number: MFCD00215198
CAS Number: 339098-46-3

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR