2-(2-Chloroacetyl)-3-(dimethylamino)acrylonitrile ≥95%

Supplier: Matrix Scientific

045587-500MG 045587-1G 045587-5G
101871-622EA 375.38 USD
101871-622 101871-624 101871-626
2-(2-Chloroacetyl)-3-(dimethylamino)acrylonitrile ≥95%
2-(2-Chloroacetyl)-3-(dimethylamino)acrylonitrile

Hazard: Irritant

Formula: C₇H₉ClN₂O
Melting Pt: 140…142 °C
MDL Number: MFCD00663746
CAS Number: 339097-02-8

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR