Methyl-3-(1-methyl-2-oxopropoxy)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045585-500MG 045585-1G 045585-5G
101871-610EA 181.54 USD
101871-610 101871-612 101871-614
Methyl-3-(1-methyl-2-oxopropoxy)-2-thiophenecarboxylate ≥95%
Methyl-3-(1-methyl-2-oxopropoxy)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₂O₄S
MW: 228.27 g/mol
Melting Pt: 54…56 °C
MDL Number: MFCD00173639
CAS Number: 339096-81-0

Order Now

Specification Test Results

Melting point 54-56°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR