2-(Aminooxy)-1-(4-phenylpiperazino)-1-ethanone hydrochloride ≥95%

Supplier: Matrix Scientific

045584-500MG 045584-1G 045584-5G
101871-604EA 375.38 USD
101871-604 101871-606 101871-608
2-(Aminooxy)-1-(4-phenylpiperazino)-1-ethanone hydrochloride ≥95%
2-(Aminooxy)-1-(4-phenylpiperazino)-1-ethanone hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃O₂
MW: 271.75 g/mol
Melting Pt: 184…188 °C
MDL Number: MFCD01114500
CAS Number: 275374-83-9

Order Now

Specification Test Results

Melting point 184-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR