2-Chloro-1-(4-phenylpiperazino)-1-ethanone ≥95%

Supplier: Matrix Scientific

045582-500MG 045582-1G 045582-5G
101871-598EA 181.54 USD
101871-598 101871-600 101871-602
2-Chloro-1-(4-phenylpiperazino)-1-ethanone ≥95%
2-Chloro-1-(4-phenylpiperazino)-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O
Melting Pt: 71…73 °C
MDL Number: MFCD00495576
CAS Number: 14761-39-8

Order Now

Specification Test Results

Melting point 71-73°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR