4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid ≥95%

Supplier: Matrix Scientific

045579-500MG 045579-1G 045579-5G
101871-580EA 747.69 USD
101871-580 101871-582 101871-584
4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid ≥95%
4-[(2-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}ethyl)amino]-4-oxo-2-butenoic acid

Hazard: Irritant

Formula: C₁₂H₁₁ClF₃N₃O₃
MW: 337.69 g/mol
Melting Pt: 166…167 °C
MDL Number: MFCD00202686
CAS Number: 339096-60-5

Order Now

Specification Test Results

Melting point 166-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR