1-[(Ammoniooxy)methyl]-2-chlorobenzene chloride ≥95%
Supplier: Matrix Scientific
045578-500MG
045578-1G
045578-5G
101871-574EA
106.15
USD
101871-574
101871-576
101871-578
1-[(Ammoniooxy)methyl]-2-chlorobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-2-chlorobenzene chloride
Hazard: Irritant
Formula:
C₇H₉Cl₂NO Melting Pt: 145-148°C |
MDL Number:
MFCD01114592 CAS Number: 5555-48-6 |
Specification Test Results
Melting point | 145-148°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...