Ethyl-4-chloro-1-(4-fluorophenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-chloro-2,3-dihydro-2-(4-fluorophenyl)pyridazin-3-one-6-carboxylate

045577-500MG 045577-1G 045577-5G
101871-568EA 430.77 USD
101871-568 101871-570 101871-572
Ethyl-4-chloro-1-(4-fluorophenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Ethyl-4-chloro-1-(4-fluorophenyl)-6-oxo-1,6-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₀ClFN₂O₃
MW: 296.69 g/mol
Melting Pt: 126…128 °C
MDL Number: MFCD00243751
CAS Number: 339031-90-2

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR