4-[(4-Methylphenyl)sulfanyl]aniline ≥95%

Supplier: Matrix Scientific

045576-500MG 045576-1G 045576-5G
101871-562EA 556.92 USD
101871-562 101871-564 101871-566
4-[(4-Methylphenyl)sulfanyl]aniline ≥95%
4-[(4-Methylphenyl)sulfanyl]aniline

Hazard: Irritant

Formula: C₁₃H₁₃NS
MW: 215.32 g/mol
Melting Pt: 73…75.5 °C
MDL Number: MFCD00172082
CAS Number: 22865-52-7

Order Now

Specification Test Results

Melting point 73-75.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR