Ethyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate ≥95%

Supplier: Matrix Scientific

045575-500MG 045575-1G 045575-5G
101871-556EA 556.92 USD
101871-556 101871-558 101871-560
Ethyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Ethyl-4-chloro-6-oxo-1-phenyl-1,6-dihydro-3-pyridazinecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O₃
MW: 278.7 g/mol
Melting Pt: 87…89.5 °C
MDL Number: MFCD00243732
CAS Number: 339030-49-8

Order Now

Specification Test Results

Melting point 87-89.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR