5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide ≥95%
Supplier: Matrix Scientific
045574-500MG
045574-1G
045574-5G
101871-550EA
216.92
USD
101871-550
101871-552
101871-554
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulfonamide ≥95%
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
Hazard: Irritant
Formula:
C₅H₈ClN₃O₂S MW: 209.66 Melting Pt: 174-176°C |
MDL Number:
MFCD00105874 CAS Number: 88398-46-3 |
Specification Test Results
Melting point | 174-176°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...