Methyl-1-(3-chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Supplier: Matrix Scientific
045573-500MG
045573-1G
045573-5G
101871-544EA
375.38
USD
101871-544
101871-546
101871-548
Methyl-1-(3-chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydro-3-pyridazinecarboxylate ≥95%
Methyl-1-(3-chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydro-3-pyridazinecarboxylate
Hazard: Irritant
Formula:
C₁₂H₉ClN₂O₄ Melting Pt: 134…136 °C |
MDL Number:
MFCD00141205 CAS Number: 339029-80-0 |
Specification Test Results
Melting point | 134-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...