1-(3-Chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid ≥97%

Supplier: Matrix Scientific

045572-500MG 045572-1G 045572-5G
101871-538EA 747.69 USD
101871-538 101871-540 101871-542
1-(3-Chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid ≥97%
1-(3-Chlorophenyl)-4-hydroxy-6-oxo-1,6-dihydro-3-pyridazinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₇ClN₂O₄
Melting Pt: 271…273 °C
MDL Number: MFCD00141204
CAS Number: 339029-56-0

Order Now

Specification Test Results

Melting point 271-273°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR