1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]piperidin-4-one

045571-500MG 045571-1G 045571-5G
101871-532EA 150.77 USD
101871-532 101871-534 101871-536
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinone ≥95%
1-(3-Chloro-5-(trifluoromethyl)-2-pyridinyl)tetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₁H₁₀ClF₃N₂O
Melting Pt: 62-65°C
MDL Number: MFCD00231641
CAS Number: 339029-35-5

Order Now

Specification Test Results

Melting point 62-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR