2-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanol ≥95%

Supplier: Matrix Scientific

045570-500MG 045570-1G 045570-5G
101871-526EA 38.46 USD
101871-526 101871-528 101871-530
2-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanol ≥95%
2-(3,5-Dimethyl-1H-pyrazol-1-yl)-1-ethanol

Hazard: Irritant

Formula: C₇H₁₂N₂O
Melting Pt: 61…64 °C
MDL Number: MFCD00141692
CAS Number: 20000-80-0

Order Now

Specification Test Results

Melting point 61-64°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR