2,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide ≥95%
Supplier: Matrix Scientific
045569-500MG
045569-1G
045569-5G
101871-520EA
181.54
USD
101871-520
101871-522
101871-524
2,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide ≥95%
2,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide
Hazard: Irritant
Formula:
C₉H₉Cl₂NO₂ Melting Pt: 128…130 °C |
MDL Number:
MFCD00141691 CAS Number: 20656-08-0 |
Specification Test Results
Melting point | 128-130°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...