5-Chloro-1-(3-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

045564-500MG 045564-1G 045564-5G
101871-500EA 186.15 USD
101871-500 101871-502 101871-504
5-Chloro-1-(3-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-1-(3-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉Cl₂NO₃
Melting Pt: 229…230.5 °C
MDL Number: MFCD00140433
CAS Number: 339024-74-7

Order Now

Specification Test Results

Melting point 229-230.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR