5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

045563-500MG 045563-1G 045563-5G
101871-494EA 556.92 USD
101871-494 101871-496 101871-498
5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-1-(4-chlorobenzyl)-6-oxo-1,6-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₉Cl₂NO₃
MW: 298.13 g/mol
Melting Pt: 230.5…231.5 °C
MDL Number: MFCD00140420
CAS Number: 339024-38-3

Order Now

Specification Test Results

Melting point 230.5-231.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR