4-Chloro-2-(4-pyridinyl)-6-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

045562-500MG 045562-1G 045562-5G
101871-488EA 375.38 USD
101871-488 101871-490 101871-492
4-Chloro-2-(4-pyridinyl)-6-(trifluoromethyl)pyrimidine ≥95%
4-Chloro-2-(4-pyridinyl)-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₅ClF₃N₃
Melting Pt: 160…161 °C
MDL Number: MFCD00140056
CAS Number: 204394-70-7

Order Now

Specification Test Results

Melting point 160-161°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR