1-Benzyl-5-chloro-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%

Supplier: Matrix Scientific

045559-500MG 045559-1G 045559-5G
101871-470EA 375.38 USD
101871-470 101871-472 101871-474
1-Benzyl-5-chloro-2-oxo-1,2-dihydro-3-pyridine carboxylic acid ≥95%
1-Benzyl-5-chloro-2-oxo-1,2-dihydro-3-pyridine carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₀ClNO₃
Melting Pt: 169…171 °C
MDL Number: MFCD00140288
CAS Number: 101384-62-7

Order Now

Specification Test Results

Melting point 169-171°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR