5-Chloro-6-oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Chloro-1,6-dihydro-6-oxo-1-[3-(trifluoromethyl)benzyl]pyridine-3-carboxylic acid
045558-500MG
045558-1G
045558-5G
101871-464EA
556.92
USD
101871-464
101871-466
101871-468
5-Chloro-6-oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid ≥95%
5-Chloro-6-oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₉ClF₃NO₃ Melting Pt: 191…193 °C |
MDL Number:
MFCD00140108 CAS Number: 339024-06-5 |
Specification Test Results
Melting point | 191-193°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...