2-(4-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-2-(pyridin-4-yl)-6-(trifluoromethyl)pyrimidine

045557-500MG 045557-1G 045557-5G
101871-458EA 278.46 USD
101871-458 101871-460 101871-462
2-(4-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol ≥95%
2-(4-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinol

Hazard: Irritant

Formula: C₁₀H₆F₃N₃O
MW: 241.17 g/mol
Melting Pt: 264…265 °C
MDL Number: MFCD00140054
CAS Number: 204394-60-5

Order Now

Specification Test Results

Melting point 264-265°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR