3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%

Supplier: Matrix Scientific

045556-500MG 045556-1G 045556-5G
101871-452EA 129.23 USD
101871-452 101871-454 101871-456
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline ≥95%
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}aniline

Hazard: Irritant

Formula: C₁₂H₈ClF₃N₂O
MW: 288.66 g/mol
Melting Pt: 66…68 °C
MDL Number: MFCD01315057
CAS Number: 80783-47-7

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR