4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

045554-500MG 045554-1G 045554-5G
101871-440EA 181.54 USD
101871-440 101871-442 101871-444
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile ≥95%
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₆ClF₃N₂O
Melting Pt: 72…74 °C
MDL Number: MFCD00170203
CAS Number: 95711-33-4

Order Now

Specification Test Results

Melting point 72-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR