2,2,2-Trifluoro-N-phenylacetamide ≥95%
Supplier: Matrix Scientific
045552-1G
045552-5G
045552-25G
101871-430EA
29.23
USD
101871-430
101871-432
101871-434
2,2,2-Trifluoro-N-phenylacetamide ≥95%
2,2,2-Trifluoro-N-phenylacetamide
Hazard: Irritant
Formula:
C₈H₆F₃NO Melting Pt: 89…91 °C |
MDL Number:
MFCD00196027 CAS Number: 404-24-0 |
Specification Test Results
Melting point | 89-91°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...