Methyl-3-(5-formyl-2-furyl)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045551-500MG 045551-1G 045551-5G
101871-424EA 278.46 USD
101871-424 101871-426 101871-428
Methyl-3-(5-formyl-2-furyl)-2-thiophenecarboxylate ≥95%
Methyl-3-(5-formyl-2-furyl)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₁H₈O₄S
MW: 236.25 g/mol
Melting Pt: 118…119 °C
MDL Number: MFCD00052169
CAS Number: 255828-29-6

Order Now

Specification Test Results

Melting point 118-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR