5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
5,7-Bis(trifluoromethyl)-1,8-naphthyridin-2-amine, 7-Amino-2,4-bis(trifluoromethyl)-1,8-naphthyridine
045550-500MG
045550-1G
045550-5G
101871-418EA
181.54
USD
101871-418
101871-420
101871-422
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-amine ≥95%
5,7-Bis(trifluoromethyl)[1,8]naphthyridin-2-amine
Hazard: Irritant
Formula:
C₁₀H₅F₆N₃ MW: 281.16 g/mol Melting Pt: 195…197 °C |
MDL Number:
MFCD00140205 CAS Number: 51420-72-5 |
Specification Test Results
Melting point | 195-197°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...