6-{[3-(Trifluoromethyl)phenyl]sulfanyl}imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

045549-500MG 045549-1G 045549-5G
101871-412EA 747.69 USD
101871-412 101871-414 101871-416
6-{[3-(Trifluoromethyl)phenyl]sulfanyl}imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%
6-{[3-(Trifluoromethyl)phenyl]sulfanyl}imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₇F₃N₂OS₂
MW: 328.34 g/mol
Melting Pt: 86…89 °C
MDL Number: MFCD00102587
CAS Number: 175277-56-2

Order Now

Specification Test Results

Melting point 86-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR