6-Chloroimidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%
Supplier: Matrix Scientific
045548-500MG
045548-1G
045548-5G
101871-406EA
747.69
USD
101871-406
101871-408
101871-410
6-Chloroimidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%
6-Chloroimidazo[2,1-b][1,3]thiazole-5-carbaldehyde
Hazard: Irritant
Formula:
C₆H₃ClN₂OS MW: 186.62 g/mol Melting Pt: 141…142 °C |
MDL Number:
MFCD00139155 CAS Number: 23576-84-3 |
Specification Test Results
Melting point | 141-142°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...