5-(3-Pyridinyl)-1,3,4-oxadiazole-2-thiol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Pyridin-3-yl)-1,3,4-oxadiazole-2-thiol

045545-500MG 045545-1G 045545-5G
101871-400EA 375.38 USD
101871-400 101871-402 101871-404
5-(3-Pyridinyl)-1,3,4-oxadiazole-2-thiol ≥95%
5-(3-Pyridinyl)-1,3,4-oxadiazole-2-thiol

Hazard: Irritant

Formula: C₇H₅N₃OS
MW: 179.2 g/mol
Melting Pt: 229…231 °C
MDL Number: MFCD00139080
CAS Number: 3690-46-8

Order Now

Specification Test Results

Melting point 229-231°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR