5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

044419-500MG 044419-1G 044419-5G
101865-736EA 747.69 USD
101865-736 101865-738 101865-740
5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%
5-[(4-Bromophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₈BrF₃N₂OS
MW: 365.18 g/mol
Melting Pt: 98…99.5 °C
MDL Number: MFCD01935942
CAS Number: 321848-45-7

Order Now

Specification Test Results

Melting point 98-99.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR