Ethyl-2,4-dimethyl-7-phenylpyrrolo[1,2-a]pyrimidine-8-carboxylate ≥95%

Supplier: Matrix Scientific

044410-500MG 044410-1G
101865-698EA 1483.08 USD
101865-698 101865-700
Ethyl-2,4-dimethyl-7-phenylpyrrolo[1,2-a]pyrimidine-8-carboxylate ≥95%
Ethyl-2,4-dimethyl-7-phenylpyrrolo[1,2-a]pyrimidine-8-carboxylate

Hazard: Irritant

Formula: C₁₈H₁₈N₂O₂
MW: 294.36 g/mol
Melting Pt: 135…138 °C
MDL Number: MFCD00171951
CAS Number: 338419-86-6

Order Now

Specification Test Results

Melting point 135-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR