1-(2-(Ammoniooxy)ethoxy)-3-(trifluoromethyl)benzene chloride ≥95%

Supplier: Matrix Scientific

Synonyms: O-{2-[3-(Trifluoromethyl)phenoxy]ethyl}hydroxylamine hydrochloride

044409-500MG 044409-1G 044409-5G
101865-692EA 747.69 USD
101865-692 101865-694 101865-696
1-(2-(Ammoniooxy)ethoxy)-3-(trifluoromethyl)benzene chloride ≥95%
1-(2-(Ammoniooxy)ethoxy)-3-(trifluoromethyl)benzene chloride

Hazard: Irritant

Formula: C₉H₁₁ClF₃NO₂
Melting Pt: 154-156°C
MDL Number: MFCD01114594
CAS Number: 113211-36-2

Order Now

Specification Test Results

Melting point 154-156°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR