1-[(Ammoniooxy)methyl]-3-fluorobenzene chloride ≥95%

Supplier: Matrix Scientific

044408-500MG 044408-1G 044408-5G
101865-686EA 155.38 USD
101865-686 101865-688 101865-690
1-[(Ammoniooxy)methyl]-3-fluorobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-3-fluorobenzene chloride

Hazard: Irritant

Formula: C₇H₉ClFNO
Melting Pt: 218-225°C
MDL Number: MFCD01114585
CAS Number: 51572-90-8

Order Now

Specification Test Results

Melting point 218-225°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR