1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride ≥95%

Supplier: Matrix Scientific

Synonyms: O-(2-Fluorobenzyl)hydroxylamine hydrochloride

044407-500MG 044407-1G 044407-5G
101865-680EA 230.77 USD
101865-680 101865-682 101865-684
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-2-fluorobenzene chloride

Hazard: Irritant

Formula: C₇H₉ClFNO
MW: 177.6039 g/mol
Melting Pt: 180-185°C
MDL Number: MFCD01114584
CAS Number: 215599-91-0

Order Now

Specification Test Results

Melting point 180-185°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR