Ethyl-3-amino-3-(4-toluidino)acrylate hydrochloride ≥95%

Supplier: Matrix Scientific

044406-500MG 044406-1G 044406-5G
101865-674EA 375.38 USD
101865-674 101865-676 101865-678
Ethyl-3-amino-3-(4-toluidino)acrylate hydrochloride ≥95%
Ethyl-3-amino-3-(4-toluidino)acrylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂
Melting Pt: 135…137 °C
MDL Number: MFCD00243955
CAS Number: 318239-42-8

Order Now

Specification Test Results

Melting point 135-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR