Ethyl-3-amino-3-(4-chloroanilino)acrylate hydrochloride ≥95%
Supplier: Matrix Scientific
044405-500MG
044405-1G
044405-5G
101865-668EA
375.38
USD
101865-668
101865-670
101865-672
Ethyl-3-amino-3-(4-chloroanilino)acrylate hydrochloride ≥95%
Ethyl-3-amino-3-(4-chloroanilino)acrylate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₄Cl₂N₂O₂ Melting Pt: 146…149 °C |
MDL Number:
MFCD00243954 CAS Number: 318239-41-7 |
Specification Test Results
Melting point | 146-149°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...