2-[(3,4-Dichlorobenzoyl)amino]acetic acid ≥95%
Supplier: Matrix Scientific
044402-500MG
044402-1G
044402-5G
101865-650EA
81.54
USD
101865-650
101865-652
101865-654
2-[(3,4-Dichlorobenzoyl)amino]acetic acid ≥95%
2-[(3,4-Dichlorobenzoyl)amino]acetic acid
Hazard: Irritant
Formula:
C₉H₇Cl₂NO₃ Melting Pt: 135…139 °C |
MDL Number:
MFCD00228361 CAS Number: 17321-80-1 |
Specification Test Results
Melting point | 135-139°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...