1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride ≥95%

Supplier: Matrix Scientific

044401-1G 044401-5G
101865-646EA 67.69 USD
101865-646 101865-648
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride ≥95%
1-[(Ammoniooxy)methyl]-4-methoxybenzene chloride

Hazard: Irritant

Formula: C₈H₁₂ClNO₂
Melting Pt: 209-211°C
MDL Number: MFCD01114582
CAS Number: 876-33-5

Order Now

Specification Test Results

Melting point 209-211°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR