1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

044398-500MG 044398-1G
101865-630EA 423.08 USD
101865-630 101865-632
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde ≥95%
1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde

Hazard: Irritant

Formula: C₉H₆ClN₃O
Melting Pt: 151…153 °C
MDL Number: MFCD01109282
CAS Number: 113934-27-3

Order Now

Specification Test Results

Melting point 151-153°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR