Methyl-2-(cyanomethoxy)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-(cyanomethoxy)benzoate
044396-500MG
044396-1G
044396-5G
101865-618EA
181.54
USD
101865-618
101865-620
101865-622
Methyl-2-(cyanomethoxy)benzenecarboxylate ≥95%
Methyl-2-(cyanomethoxy)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₀H₉NO₃ MW: 191.19 g/mol Melting Pt: 53…55 °C |
MDL Number:
MFCD01924694 CAS Number: 1641-00-5 |
Specification Test Results
Melting point | 53-55°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...