2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinethiol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Pyridin-2-yl)-4-thio-6-(trifluoromethyl)pyrimidine
044393-500MG
044393-1G
044393-5G
101865-600EA
921.54
USD
101865-600
101865-602
101865-604
2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinethiol ≥95%
2-(2-Pyridinyl)-6-(trifluoromethyl)-4-pyrimidinethiol
Hazard: Irritant
Formula:
C₁₀H₆F₃N₃S MW: 257.24 g/mol Melting Pt: 145…148 °C |
MDL Number:
MFCD00139802 CAS Number: 338418-16-9 |
Specification Test Results
Melting point | 145-148°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...