Methyl-2-[(4-benzyl-1-phthalazinyl)sulfanyl]acetate ≥95%

Supplier: Matrix Scientific

044392-500MG 044392-1G 044392-5G
101865-594EA 747.69 USD
101865-594 101865-596 101865-598
Methyl-2-[(4-benzyl-1-phthalazinyl)sulfanyl]acetate ≥95%
Methyl-2-[(4-benzyl-1-phthalazinyl)sulfanyl]acetate

Hazard: Irritant

Formula: C₁₈H₁₆N₂O₂S
Melting Pt: 120…122 °C
MDL Number: MFCD00139962
CAS Number: 338417-99-5

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR