Methyl-3-[(4-fluorobenzyl)oxy]-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
044391-500MG
044391-1G
044391-5G
101865-588EA
181.54
USD
101865-588
101865-590
101865-592
Methyl-3-[(4-fluorobenzyl)oxy]-2-thiophenecarboxylate ≥95%
Methyl-3-[(4-fluorobenzyl)oxy]-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₃H₁₁FO₃S Melting Pt: 90…91 °C |
MDL Number:
MFCD00138942 CAS Number: 338417-95-1 |
Specification Test Results
Melting point | 90-91°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...