3-Methyl-4-[(4-methylphenyl)sulfanyl]-1H-pyrazole ≥95%

Supplier: Matrix Scientific

044390-500MG 044390-1G 044390-5G
101865-582EA 375.38 USD
101865-582 101865-584 101865-586
3-Methyl-4-[(4-methylphenyl)sulfanyl]-1H-pyrazole ≥95%
3-Methyl-4-[(4-methylphenyl)sulfanyl]-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₁₂N₂S
Melting Pt: 120…122 °C
MDL Number: MFCD00138929
CAS Number: 318238-20-9

Order Now

Specification Test Results

Melting point 120-122°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR