Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

044388-500MG 044388-1G 044388-5G
101865-570EA 747.69 USD
101865-570 101865-572 101865-574
Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate ≥95%
Methyl-3-(2-chloro-6-fluorophenyl)-5-[2-(dimethylamino)vinyl]-4-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₄ClFN₂O₃
Melting Pt: 139…141 °C
MDL Number: MFCD00127467
CAS Number: 338417-50-8

Order Now

Specification Test Results

Melting point 139-141°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR