Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

044387-500MG 044387-1G 044387-5G
101865-564EA 375.38 USD
101865-564 101865-566 101865-568
Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate ≥95%
Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₂H₉ClFNO₃
MW: 269.66 g/mol
Melting Pt: 53…55 °C
MDL Number: MFCD00214743
CAS Number: 4415-09-2

Order Now

Specification Test Results

Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR