Ethyl-2-{2-methoxy-6-[(methoxyimino)methyl]phenoxy}acetate ≥95%

Supplier: Matrix Scientific

044384-500MG 044384-1G 044384-5G
101865-556EA 375.38 USD
101865-556 101865-558 101865-560
Ethyl-2-{2-methoxy-6-[(methoxyimino)methyl]phenoxy}acetate ≥95%
Ethyl-2-{2-methoxy-6-[(methoxyimino)methyl]phenoxy}acetate

Hazard: Irritant

Formula: C₁₃H₁₇NO₅
Melting Pt: 51…53 °C
MDL Number: MFCD00127398
CAS Number: 338416-55-0

Order Now

Specification Test Results

Melting point 51-53°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR